Learn More
Fisherbrand™ Delrin™ Half-Size Test Tube Racks, 36 x 13mm
GREENER_CHOICE
Product Code. 15310410 Shop All Fisherbrand Products
Click to view available options
Color:
Blue
Green
Magenta
White
Unit Size:
Each
Product Code. 15310410
Only null left
Add to basket
Add to basket
Molded in a single, continuous piece. Fisherbrand™ Delrin™ Half-Size Test Tube Racks, 36 x 13mm are highly resistant to heat and chemical attack.
  • Delrin™ fiberglass-reinforced polyoxymethylene (POM)
  • Offers the highest chemical and heat resistance (-40 to +140°C)
  • Autoclavable
  • Molded coordinates for tube orientation
  • Stable design for use in water baths
  • Flat labeling area for easier identification
Specifications
Product Type Test Tube Rack
Array 6 x 6
Autoclavable Autoclavable
Color Green
Holds Tubes (13 mm), Tubes (5 to 10 mL)
Material Fiberglass-reinforced POM
Positions 36
Shape Teardrop-shaped Openings
Shelves 3 Tiers
Stackable No
Sterility Non-sterile
For Use With (Equipment) 5 to 10mL test tubes
Quantity Each
Dimensions (L x W x H) 104 x 104 x 59mm
Length (Metric) 104 mm
Width (Metric) 104 mm
Height (Metric) 59 mm
Diameter (Metric) Opening 13 mm