missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fetuin-A Rabbit anti-Human, Clone: 10000, Proteintech
Rabbit Recombinant Antibody
1757.51 NOK - 5178.17 NOK
Specifications
| Antigen | Fetuin-A |
|---|---|
| Clone | 10000 |
| Concentration | 500 μg/mL |
| Applications | Western Blot |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17649796
|
Proteintech
80793-1-RR-20UL |
20 μL |
1757.51 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17639796
|
Proteintech
80793-1-RR-100UL |
100 μL |
5178.17 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fetuin-A antibody apecifically detetects Fetuin-A antigen in Human Samples. It is validated for Western Blot.Specifications
| Fetuin-A | |
| 500 μg/mL | |
| Unconjugated | |
| Rabbit | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| 197 | |
| Fetuin-A fusion protein Ag9853 | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 10000 | |
| Western Blot | |
| Liquid | |
| Human | |
| BC048198 | |
| AHSG | |
| IgG | |
| Protein A purification |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title