missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fetuin-A Rabbit anti-Human, Clone: 10000, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 80793-1-RR-20UL
This item is not returnable.
View return policy
Description
Fetuin-A antibody apecifically detetects Fetuin-A antigen in Human Samples. It is validated for Western Blot.Specifications
| Fetuin-A | |
| 10000 | |
| Unconjugated | |
| BC048198 | |
| Rabbit | |
| Protein A purification | |
| Primary | |
| Human | |
| Liquid |
| Western Blot | |
| 500 μg/mL | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| AHSG | |
| Fetuin-A fusion protein Ag9853 | |
| 20 μL | |
| 197 | |
| Store at -20°C. Stable for one year after shipment. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction