Learn More
Ferrocenecarboxylic Acid, TRC
Product Code. 30400918
Change view
Click to view available options
Quantity:
2.5 g
5 g
25 g
Unit Size:
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400918 2.5 g 2.5g
30408041 5 g 5g
30406410 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30400918 Supplier Toronto Research Chemicals Supplier No. TRCF3076182.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1271-42-7
Molecular Formula C11H10FeO2
Molecular Weight (g/mol) 230.04
Synonym Iron, (carboxycyclopentadienyl)cyclopentadienyl- (6CI), Carboxyferrocene
IUPAC Name Ferrocenemonocarboxylic acid
SMILES O=C(O)[C-]12[CH]3=[CH]4[CH]5=[CH]1[Fe+2]67894532[CH]=%10[CH]9=[CH]8[CH-]7[CH]%106
Specifications
Quantity 2.5 g
InChI Formula InChI=1S/C6H5O2.C5H5.Fe/c7-6(8)5-3-1-2-4-5;1-2-4-5-3-1;/h1-4H,(H,7,8);1-5H;/q2*-1;+2
Formula Weight 230
Chemical Name or Material Ferrocenecarboxylic Acid