accessibility menu, dialog, popup

UserName

CAS RN 1271-42-7

FeOOH

CAS RN 1271-42-7

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 225.01
Molecular Formula: C11H5FeO2
InChi Key: OUPWBVGTMCICLR-UHFFFAOYSA-N
SMILES: [C-]12[C-]3[C-]4[C-]5[C-]1[Fe]2345.OC(=O)[c-]1cccc1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results