Learn More
Thermo Scientific™ DEPC-treated Water
Product Code. p-4530715 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
30 mL
5 x 1 mL
Unit Size:
30mL
Pack of 5
Product Code. Quantity unitSize
11531575 5 x 1 mL Pack of 5
10213130 30 mL 30mL
2 options
Product Code. 11531575
Brand: Thermo Scientific™ R0603
Optimize applications involving RNA with this deionized, diethylpyrocarbonate (DEPC) treated and 0.22μm membrane-filtered water.
deionized, diethylpyrocarbonate (DEPC) is treated and 0.22 μm membrane-filtered water. It is ideal for applications involving RNA.
Ideal for applications involving RNA.
  • Free of endo-, exodeoxyribonucleases, ribonucleases and phosphatases
Note:
DEPC-treated Water is not recommended for PCR or in vivo experiments (1).
TRUSTED_SUSTAINABILITY
Specifications
pH 7.9
Grade Molecular Biology
Packaging Vial
Treatment(s) DEPC-Treated
Quantity 5 x 1 mL