Learn More
FBXO32 Mouse anti-Human, Unconjugated, Clone: 3C2G3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67172-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The FBXO32 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FBXO32. This antibody recognizes human, pig, rat, mouse antigen. The FBXO32 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
FBXO32 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |
Western Blot, Immunohistochemistry | |
3C2G3 | |
Unconjugated | |
150 μL | |
114907 | |
IgG2b |