missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO32 Mouse anti-Human, Unconjugated, Clone: 3C2G3, Proteintech
Mouse Monoclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | FBXO32 |
---|---|
Clone | 3C2G3 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16889655
|
Proteintech
67172-1-IG-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16879655
|
Proteintech
67172-1-IG-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The FBXO32 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FBXO32. This antibody recognizes human, pig, rat, mouse antigen. The FBXO32 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
FBXO32 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
114907 | |
Primary |
3C2G3 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Human, Mouse, Rat, Pig | |
IgG2b |