Learn More
FBL Mouse anti-Human, Unconjugated, Clone: 3A9E8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66985-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16889325
  • 1611.84 NOK / 20µL
Estimated Shipment: 16-05-2024
Description
The FBL antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FBL. This antibody recognizes human, rat, mouse antigen. The FBL antibody has been validated for the following applications: FC, IP, WB, ELISA analysis.
Specifications
Specifications
FBL
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunoprecipitation, Flow Cytometry
3A9E8
Unconjugated
20 μL
2091
IgG1