missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBL Mouse anti-Human, Unconjugated, Clone: 3A9E8, Proteintech
Mouse Monoclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | FBL |
---|---|
Clone | 3A9E8 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunoprecipitation, Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16889325
|
Proteintech
66985-1-IG-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16879325
|
Proteintech
66985-1-IG-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The FBL antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FBL. This antibody recognizes human, rat, mouse antigen. The FBL antibody has been validated for the following applications: FC, IP, WB, ELISA analysis.
Specifications
FBL | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
2091 | |
Primary |
3A9E8 | |
Western Blot, Immunoprecipitation, Flow Cytometry | |
Unconjugated | |
Human, Rat, Mouse | |
IgG1 |