Learn More
FBL Mouse anti-Human, Unconjugated, Clone: 3A9E8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66985-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The FBL antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FBL. This antibody recognizes human, rat, mouse antigen. The FBL antibody has been validated for the following applications: FC, IP, WB, ELISA analysis.
Specifications
FBL | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunoprecipitation, Flow Cytometry | |
3A9E8 | |
Unconjugated | |
20 μL | |
2091 | |
IgG1 |