Description
- Polycarbonate construction and available in three colors for user/owner identification
- Silicone water-tight seal and three lid clasps provide secure closure
- Carry handle folds neatly into body space allowing the units to be stacked
- Lid opens a full 180°, allowing total access to contents and making it easier to clean
- Accommodates 13 and 16mm tubes in a 72-place onerack™, or similar sized/brand of rack
- Separators provided to create own compartment sizes, or use empty to transport larger products and equipment
- Autoclavable
Specifications
Specifications
| Product Type | Box |
| Color | Yellow |
| Material | Polycarbonate |
| Dimensions (L x W x H) | 15.25 x 8.25 x 6.75 in. (38.73 x 20.95 x 17.14 cm) |
| For Use With (Application) | Transporting |
| For Use With (Equipment) | 13 and 16mm tubes in a 72-place rack |
| Height (English) | 6.75 in. |
| Height (Metric) | 17.14 cm |
| Holds | 13 to 16 mm Tubes in a 72-Place Rack |
| Includes | (3) Lid Clasps, Separator, Water-Tight Seal |
| Show More |