Description
- Adaptors for rotors
- Available in packs of 6
Specifications
Specifications
| Type | Adapter |
| Tube Type | PCR |
| Capacity | 6 x 0.2 mL PCR Tubes |
| For Use With (Equipment) | Rotor F-45-48-11 |
| Angle | Fixed |
| For Use With (Application) | 0.2mL PCR Tubes |
| No. per Pack | 6/Pk. |