missing translation for 'onlineSavingsMsg'
Learn More

FAM168B Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 24422-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16892895

  • 4593.76 NOK / 150µL
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Modulates neuronal axonal outgrowth by acting as a negative regulator of CDC42 and STAT3 and a positive regulator of STMN2. Positive regulator of CDC27.
Specifications

Specifications

FAM168B
Polyclonal
Unconjugated
FAM168B
FAM168B, KIAA0280L, p20, Protein FAM168B
Rabbit
Antigen Affinity Chromatography
RUO
130074
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.44 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
A1KXE4
FAM168B
FAM168B Fusion Protein Ag19535
150 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers