missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM168B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | FAM168B |
---|---|
Concentration | 0.44 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16802905
|
Proteintech
24422-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16892895
|
Proteintech
24422-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Modulates neuronal axonal outgrowth by acting as a negative regulator of CDC42 and STAT3 and a positive regulator of STMN2. Positive regulator of CDC27.Specifications
FAM168B | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
A1KXE4 | |
130074 | |
FAM168B Fusion Protein Ag19535 | |
Primary | |
-20°C | |
FAM168B |
0.44 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
FAM168B, KIAA0280L, p20, Protein FAM168B | |
FAM168B | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |