missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM168B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24422-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Modulates neuronal axonal outgrowth by acting as a negative regulator of CDC42 and STAT3 and a positive regulator of STMN2. Positive regulator of CDC27.Specifications
FAM168B | |
Polyclonal | |
Unconjugated | |
FAM168B | |
FAM168B, KIAA0280L, p20, Protein FAM168B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
130074 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.44 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A1KXE4 | |
FAM168B | |
FAM168B Fusion Protein Ag19535 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |