Learn More
IBA Lifesciences FABian™ Complete System
Product Code. 15839814 Shop All IBA Lifesciences Products
Click to view available options
Quantity:
1
Unit Size:
Each
Product Code. 15839814
Brand: IBA Lifesciences 66100260
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
FABian Complete System is for reversible cell isolation delivering label-free target cells.
  • The FABian™ cell selection system is a fully automatic bench top instrument, which utilizes IBA’s reversible Fab column-based Traceless Affinity Cell Selection (Fab-TACS™) procedure for non-magnetic isolation of PBMCs, T, B and other cells of interest with high efficiency and purity directly from whole blood, buffy coat or other single-cell tissue suspensions.
Specifications
Includes Automated cell selection system with 10 mL syringe adapter
For Use With (Application) PBMC, T and B Cell Isolation
Quantity 1