Learn More
Gilson™ Extraction Cartridge Cap
Product Code. 16686155 Shop All Gilson Products
Change view
Click to view available options
Volume (Metric):
1 mL
3 mL
6 mL
8 mL
Unit Size:
Pack of 1000
Pack of 250
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
16686155 8 mL Pack of 250
16656155 1 mL Pack of 1000
16666155 3 mL Pack of 1000
16676155 6 mL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16686155 Supplier Gilson™ Supplier No. 2954731
  • ONLINE EXCLUSIVE PRICE
    3230.00 NOK
    2500.62 NOK / Pack of 250
    Save 729.38 NOK
    22% Off
 Restricted Item
In Stock
Gilson offers a variety of vials and caps in bulk for a number of applications.
  • ASPEC Caps
  • Gilson offers a variety of vials and caps in bulk for a number of applications.
Specifications
Description ASPEC Caps
Volume (Metric) 8 mL
Material Glass Decs
Quantity 250 Pk.