Learn More
Memmert™ Extended Overtemperature Protection for SN55
Extended overtemperature protection with integrated Pt100 sensor for independent temperature monitoring.
Brand:  Memmert™ SN55A6
Additional Details : Weight : 0.01200kg
Product Code. 17508665
  • 2065.00 NOK / Each
Estimated Shipment: 20-06-2024
Description
  • Accessory
  • With integrated Pt100 sensor
  • For models with SingleDISPLAY
Specifications
Specifications
SN55 Sterilizer
Integrated Pt100 Sensor