Learn More
Memmert™ Extended Overtemperature Protection for SF450
Product Code. 17598255 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
SF450 Sterilizer
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
17598255 SF450 Sterilizer Each
1 options
Product Code. 17598255
Brand: Memmert™ SF450A6
Only null left
Add to basket
Add to basket
Extended overtemperature protection with integrated Pt100 sensor for independent temperature monitoring.
  • Accessory
  • With integrated Pt100 sensor
  • For models with SingleDISPLAY
Specifications
For Use With (Equipment) SF450 Sterilizer
Includes Integrated Pt100 Sensor