Learn More
BINDER™ Calibration Certificate, Expanded
Shop All BINDER Products
Click to view available options
For Use With (Equipment):
BINDER Chambers Series MK
BINDER Chambers Series MKF
BINDER Chambers Series MKFT
BINDER Chambers Series MKT
Unit Size:
Each
Description
- Each additional measurement at an additional measuring point or set of values
- Extension
Specifications
Specifications
| For Use With (Equipment) | BINDER Chambers Series MKFT |
| Description | Calibration certificate, expanded - for temperature and humidity; for extending the measurement in center of chamber to include another test value |
| Calibration Points | 1 Measuring Point |
| Duration | 60 to 120 min. |
| Calibration Type | Temperature and Humidity Measurement |
| Product Type | Calibration Certificate (Expanded) |