Learn More
Eppendorf™ Exchangeable Thermoblock
Product Code. 10344651 Shop All Eppendorf Products
Change view
Click to view available options
Holds:
0.5 mL Tubes
1.5 mL tubes
Unit Size:
Each
Product Code. Holds unitSize
10344651 0.5 mL Tubes Each
10439472 1.5 mL tubes Each
2 options
Product Code. 10344651
Brand: Eppendorf™ 5361000015
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Accessories for ThermoStat plus/thermoMixer plus
  • Compact, bench top thermostat with thermoblocks for 0.2mL, 0.5mL, 1.0mL, 2.0mL tubes and microtitre plates designed to provide a high degree of flexibility for a multitude of applications
  • Programmable temperature and run time for special applications
  • GLP compliant
  • RS232 interface
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 24 Reaction Vessels 0.5mL with Work Rack and Cool Pack 0°C
Holds 0.5 mL Tubes
No. of Places 24
Type Block