Learn More
Thermo Scientific™ Eutech™ Salinity (NaCl) Standard Solutions
Product Code. p-7114270
Change view
Click to view available options
Concentration:
25 ppt
5 ppt
Unit Size:
480mL
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Concentration unitSize
11579124 25 ppt 480mL
15601717 5 ppt Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11579124 Supplier Thermo Scientific™ Supplier No. 01X211231
Only null left
Add to basket
Add to basket
Eutech™ offers Salinity Standard Solutions for use with Salinity Meters.
Eutech™ offers a range of Salinity Standard Solutions for use with Salinity Meters. Salinity measurements are common in industries like agriculture, aquaculture, hydroponics, food, pools and spas where it is necessary to monitor the salt level constantly.
Features:
  • Buffers and calibration solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.
Description:
  • ECNACL5PPT: 5 ppt (parts per thousand) Nacl calibration solution
  • ECNACL25PPT: 25 ppt (parts per thousand) Nacl calibration solution
  • ECNACL45PPT: 45 ppt(parts per thousand) Nacl calibration solution
TRUSTED_SUSTAINABILITY
Specifications
Concentration 25 ppt
Description 25 ppt NaCl Calibration Solution
Type NaCl Calibration Solution