Learn More
Ethyl Acetate, Optima™ for HPLC and GC, Thermo Scientific™
Product Code. 15899164 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Amber Soda Lime Glass
Unit Size:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15899164 4 L Amber Soda Lime Glass 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15899164 Supplier Fisher Chemical Supplier No. E1964
  • ONLINE EXCLUSIVE PRICE
    24715.00 NOK
    23991.79 NOK / 4 L
    Save 723.21 NOK
    2% Off
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
High purity and versatile solvent suitable for HPLC, spectrophotometry, and pesticide residue analysis
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Tested to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
  • Also meets ACS specifications
  • Intended for research use only
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C
Specifications
Melting Point -83.5°C
Absorbance 0.015 AU max. at 270nm, 0.12 AU max. at 260nm, 1.00 AU max. at 255nm
Color Colorless
Density 0.902g/cm³
Quantity 4 L
Identification Pass Test
Packaging Amber Soda Lime Glass
Physical Form Liquid
Refractive Index 1.3680 to 1.3710 (at 25°C)
Residue after Evaporation 1ppm max.
Assay Percent Range Assay: 99.9% min by GC
Titratable Acid 0.0009mEq/g max.
Linear Formula CH3COOC2H5
Vapor Pressure 103mbar at 20°C
Viscosity 0.45 cP at 20°C
Water 0.02% max.
Boiling Point 75°C
Formula Weight 88.11
Percent Purity ≥99.9%
Grade High purity mobile phase for HPLC and/or extraction solvent for GC applications
Chemical Name or Material Ethyl Acetate