missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-fluorobenzo[b]thiophene-2-carboxylate, 96%, Thermo Scientific™

Product Code. 15497097
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
15497097 5 g 5g
15458805 250 mg 250mg
15487097 1 g 1g
3 options
This item is not returnable. View return policy

Product Code. 15497097

Brand: Thermo Scientific Alfa Aesar H34167.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13771-69-2
Molecular Formula C11H9FO2S
Molecular Weight (g/mol) 224.249
MDL Number MFCD12923024
InChI Key PQGYDIBMOHLOAA-UHFFFAOYSA-N
Synonym ethyl 5-fluorobenzo b thiophene-2-carboxylate, benzo b thiophene-2-carboxylic acid, 5-fluoro-, ethyl ester
PubChem CID 23261724
IUPAC Name ethyl 5-fluoro-1-benzothiophene-2-carboxylate
SMILES CCOC(=O)C1=CC2=C(S1)C=CC(=C2)F

Specifications

Melting Point 88°C to 89°C
Quantity 5 g
Formula Weight 224.26
Percent Purity 96%
Chemical Name or Material Ethyl 5-fluorobenzo[b]thiophene-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.