accessibility menu, dialog, popup

UserName

CAS RN 13771-69-2

H3COOSF

CAS RN 13771-69-2

IUPAC Name: ethyl 5-fluoro-1-benzothiophene-2-carboxylate
Synonyms: ethyl 5-fluoro-1-benzothiophene-2-carboxylate
Molecular Weight (g/mol): 224.25
Molecular Formula: C11H9FO2S
InChi Key: PQGYDIBMOHLOAA-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC2=CC(F)=CC=C2S1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results