missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-methyl-3-phenylglycidate, cis + trans, 98%

Product Code. 11490048
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490048 250 g 250g
11480048 50 g 50g
11470048 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490048 Supplier Thermo Scientific Alfa Aesar Supplier No. B23654.30
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 77-83-8 | C12H14O3 | 206.241 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-83-8
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.241
MDL Number MFCD00022338
InChI Key LQKRYVGRPXFFAV-UHFFFAOYSA-N
Synonym ethyl methylphenylglycidate, ethyl 3-methyl-3-phenylglycidate, strawberry aldehyde, fraeseol, empg, c-16 aldehyde, aldehyde c-16, ethyl methylphenyl glycidate, ethyl-3-methyl-3-phenylglycidate, ethyl 2,3-epoxy-3-phenylbutyrate
PubChem CID 6501
IUPAC Name ethyl 3-methyl-3-phenyloxirane-2-carboxylate
SMILES CCOC(=O)C1C(O1)(C)C2=CC=CC=C2

Specifications

Density 1.044
Boiling Point 272°C to 275°C
Flash Point 134°C (273°F)
Refractive Index 1.505
Quantity 250 g
Beilstein 12299
Formula Weight 206.24
Percent Purity 98%
Chemical Name or Material Ethyl 3-methyl-3-phenylglycidate, cis + trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.