accessibility menu, dialog, popup

UserName

CH3OOOH3C

CAS RN 77-83-8

IUPAC Name: ethyl 3-methyl-3-phenyloxirane-2-carboxylate
Synonyms: ethyl methylphenylglycidate
Molecular Weight (g/mol): 206.24
Molecular Formula: C12H14O3
InChi Key: LQKRYVGRPXFFAV-UHFFFAOYNA-N
SMILES: CCOC(=O)C1OC1(C)C1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)