Learn More
Ethyl 3,5-bis(trifluoromethyl)benzoate, 99%, Thermo Scientific™
Product Code. 11456826
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 11456826
Brand: Thermo Scientific Alfa Aesar B23952.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 96617-71-9
Molecular Formula C11H8F6O2
Molecular Weight (g/mol) 286.17
MDL Number MFCD01320684
InChI Key NBWZJMOEFTYMOF-UHFFFAOYSA-N
Synonym ethyl 3,5-bis trifluoromethyl benzoate, 3,5-bis-trifluoromethyl-benzoic acid ethyl ester, 3,5-bis trifluoromethyl benzoic acid ethyl ester, rarechem al bi 0207, acmc-20al07, ethyl 3?5-bis trifluoromethyl benzoate, ethyl 3 5-bis trifluoromethyl benzoate, benzoic acid, 3,5-bis trifluoromethyl-, ethyl ester
PubChem CID 2733458
IUPAC Name ethyl 3,5-bis(trifluoromethyl)benzoate
SMILES CCOC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
Specifications
Quantity 1 g
Formula Weight 286.17
Percent Purity 99%
Chemical Name or Material Ethyl 3,5-bis(trifluoromethyl)benzoate