CAS RN 96617-71-9
IUPAC Name:
ethyl 3,5-bis(trifluoromethyl)benzoate
Synonyms:
ethyl 3,5-bis(trifluoromethyl)benzoate
Molecular Weight (g/mol):
286.17
Molecular Formula:
C11H8F6O2
InChi Key:
NBWZJMOEFTYMOF-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
1
–
2
of
2
results
2
Ethyl 3,5-bis(trifluoromethyl)benzoate, 99%, Thermo Scientific™
CAS: 96617-71-9 Molecular Formula: C11H8F6O2 Molecular Weight (g/mol): 286.17 MDL Number: MFCD01320684 InChI Key: NBWZJMOEFTYMOF-UHFFFAOYSA-N Synonym: ethyl 3,5-bis trifluoromethyl benzoate,3,5-bis-trifluoromethyl-benzoic acid ethyl ester,3,5-bis trifluoromethyl benzoic acid ethyl ester,rarechem al bi 0207,acmc-20al07,ethyl 3?5-bis trifluoromethyl benzoate,ethyl 3 5-bis trifluoromethyl benzoate,benzoic acid, 3,5-bis trifluoromethyl-, ethyl ester PubChem CID: 2733458 IUPAC Name: ethyl 3,5-bis(trifluoromethyl)benzoate SMILES: CCOC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F