missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2,4-dimethylnicotinate, 96%

Product Code. 11372646
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11372646 5 g 5g
11362646 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372646 Supplier Thermo Scientific Alfa Aesar Supplier No. L08672.06
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 37669-78-6 | C10H13NO2 | 179.219 g/mol

It is employed in the design, synthesis, and evaluation of a family of propargyl pyridinyl ethers as potential cytochrome p450 inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the design, synthesis, and evaluation of a family of propargyl pyridinyl ethers as potential cytochrome p450 inhibitors.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37669-78-6
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00051962
InChI Key DIZHKKYVLBSXAW-UHFFFAOYSA-N
Synonym ethyl 2,4-dimethylnicotinate, ethyl-2,4-dimethylpyridine-3-carboxylate, 3-pyridinecarboxylic acid, 2,4-dimethyl-, ethyl ester, 2,4-dimethyl-3-pyridinecarboxlic acid ethyl ester, ethyl2,4-dimethylnicotinate, ethyl 2,4-dimethyl nicotinate, 2,4-dimethyl-3-carboethoxypyridine, 2,4-dimethyl-nicotinic acid ethyl ester, ethyl 2,4-dimethyl-3-pyridinecarboxylate, 2,4-dimethyl-3-pyridinecarboxic acid ethyl ester
PubChem CID 736245
IUPAC Name ethyl 2,4-dimethylpyridine-3-carboxylate
SMILES CCOC(=O)C1=C(C=CN=C1C)C

Specifications

Boiling Point 116°C to 118°C (10 mmHg)
Refractive Index 1.498
Quantity 5 g
Beilstein 133910
Solubility Information Slightly soluble in water.
Formula Weight 179.22
Percent Purity 96%
Chemical Name or Material Ethyl 2,4-dimethylnicotinate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.