accessibility menu, dialog, popup

UserName

H3COOCH3NH3C

CAS RN 37669-78-6

IUPAC Name: ethyl 2,4-dimethylpyridine-3-carboxylate
Synonyms: ethyl 2,4-dimethylpyridine-3-carboxylate
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: DIZHKKYVLBSXAW-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)C=CN=C1C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results