Learn More
Memmert™ Ethernet Connection Cable
Product Code. 17110065 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP1400eco
HPP2200eco
HPP260eco
HPP410eco
HPP750eco
ICO105
ICO150
ICO240
ICO50
IPP1060eco
IPP110eco
IPP1400eco
IPP2200eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
17110065 IPP1060eco Each
17100035 ICO50 Each
17100105 IPP110eco Each
17109934 ICO105 Each
17129724 HPP1400eco Each
17130135 IPP1400eco Each
17130225 IPP410eco Each
17139644 HPP1060eco Each
17139874 HPP750eco Each
17139964 ICO150 Each
17140155 IPP2200eco Each
17140185 IPP260eco Each
17149824 HPP410eco Each
17160265 IPP750eco Each
17169684 HPP110eco Each
17169744 HPP2200eco Each
17179994 ICO240 Each
17199774 HPP260eco Each
18 options
Product Code. 17110065
Brand: Memmert™ IPP1060ecoE06189
Only null left
Add to basket
Add to basket
5 m ethernet connection cable for computer interface
  • For computer interface
  • Accessory
  • Notification by SMS in case of any error or alarm of the device
Specifications
For Use With (Equipment) IPP1060eco
Length (Metric) 5 m