missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESYT2 Mouse, anti-Human, Clone: 1B12G2, Proteintech
Mouse Monoclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | ESYT2 |
|---|---|
| Clone | 1B12G2 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18069865
|
Proteintech
68374-1-IG-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18038195
|
Proteintech
68374-1-IG-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ESYT2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ESYT2. This antibody recognizes human antigen. The ESYT2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ESYT2 | |
| 1000 μg/mL | |
| Liquid | |
| Human | |
| CHR2SYT, E Syt2, ESYT2, Extended synaptotagmin 2, FAM62B, KIAA1228 | |
| Primary |
| 1B12G2 | |
| Western Blot, ELISA | |
| Mouse | |
| 57488 | |
| IgG1 | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title