Learn More
ESYT2 Mouse, anti-Human, Clone: 1B12G2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68374-1-IG-150UL
 View more versions of this product
Product Code. 18069865
  • 4836.10 NOK / 150µL
Description
The ESYT2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ESYT2. This antibody recognizes human antigen. The ESYT2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
ESYT2
1B12G2
CHR2SYT, E Syt2, ESYT2, Extended synaptotagmin 2, FAM62B, KIAA1228
150 μL
57488
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
1000 μg/mL
Mouse
Primary
Human
150 kDa
IgG1