missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERp57/ERp60 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | ERp57/ERp60 |
|---|---|
| Concentration | 0.17 mg/mL |
| Applications | Immunofluorescence, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16855384
|
Proteintech
15967-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16845384
|
Proteintech
15967-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a protein of the endoplasmic reticulum that interacts with lectin chaperones calreticulin and calnexin to modulate folding of newly synthesized glycoproteins. The protein was once thought to be a phospholipase; however, it has been demonstrated that the protein actually has protein disulfide isomerase activity. It is thought that complexes of lectins and this protein mediate protein folding by promoting formation of disulfide bonds in their glycoprotein substrates.Specifications
| ERp57/ERp60 | |
| Immunofluorescence, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P11598, P27773, P30101 | |
| 14827, 2923, 29468 | |
| ERp57/ERp60 Fusion Protein Ag8741 | |
| Primary | |
| -20°C | |
| PDIA3 |
| 0.17 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| 58 kDa microsomal protein, Disulfide isomerase ER 60, ER protein 57, ER protein 60, ER60, ERp57, ERp60, ERp61, GRP57, GRP58, HsT17083, P58, PDIA3, PI PLC, Protein disulfide isomerase A3 | |
| PDIA3 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title