missing translation for 'onlineSavingsMsg'
Learn More

ERp57/ERp60 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16855384 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16855384 20 μL 20µL
16845384 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16855384

Brand: Proteintech 159671AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes a protein of the endoplasmic reticulum that interacts with lectin chaperones calreticulin and calnexin to modulate folding of newly synthesized glycoproteins. The protein was once thought to be a phospholipase; however, it has been demonstrated that the protein actually has protein disulfide isomerase activity. It is thought that complexes of lectins and this protein mediate protein folding by promoting formation of disulfide bonds in their glycoprotein substrates.

Specifications

Antigen ERp57/ERp60
Applications Immunofluorescence, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry
Classification Polyclonal
Concentration 0.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PDIA3
Gene Accession No. P11598, P27773, P30101
Gene Alias 58 kDa microsomal protein, Disulfide isomerase ER 60, ER protein 57, ER protein 60, ER60, ERp57, ERp60, ERp61, GRP57, GRP58, HsT17083, P58, PDIA3, PI PLC, Protein disulfide isomerase A3
Gene Symbols PDIA3
Host Species Rabbit
Immunogen ERp57/ERp60 Fusion Protein Ag8741
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14827, 2923, 29468
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.