Learn More
Eppendorf™ epT.I.P.S.™ Standard Pipette Tips
Product Code. p-8000185 Shop All Eppendorf Products
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL
0.1 to 20 μL
0.5 to 10 mL
0.5 to 20 μL
100 to 2500 μL
100 to 5000 μL
1000 to 10,000 μL
2 to 200 μL
20 to 300 μL
50 to 1000 μL
50 to 1250 μL
Length (Metric):
34 mm
40 mm
46 mm
53 mm
55 mm
71 mm
76 mm
103 mm
115 mm
120 mm
165 mm
243 mm
Color Code:
Blue
Dark Gray
Dark Green
Green
Light Gray
Medium Gray
Orange
Red
Turquoise
Violet
Yellow
Color:
Blue
Colorless
Yellow
Unit Size:
Pack of 1000
Pack of 200
Pack of 500
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Length (Metric) Color Code Color Unit Size
10708972 0.5 to 20 μL 46 mm Light Gray Colorless Pack of 1000
10157291 100 to 5000 μL 120 mm Violet Colorless Pack of 500
10333682 0.5 to 10 mL 165 mm Turquoise Colorless Pack of 200
10199171 1000 to 10,000 μL 243 mm Turquoise Colorless Pack of 200
11920537 2 to 200 μL 53 mm Yellow Colorless Pack of 1000
10248621 2 to 200 μL 53 mm Yellow Yellow Pack of 1000
10177241 20 to 300 μL 55 mm Orange Yellow Pack of 1000
10370842 50 to 1000 μL 71 mm Blue Blue Pack of 1000
10772559 50 to 1000 μL 71 mm Blue Colorless Pack of 1000
10218671 100 to 2500 μL 115 mm Red Colorless Pack of 500
10197241 50 to 1250 μL 76 mm Green Colorless Pack of 1000
10788962 0.1 to 10 μL 34 mm Dark Gray Colorless Pack of 1000
10426461 0.1 to 20 μL 40 mm Medium Gray Colorless Pack of 1000
10744587 50 to 1250 μL 103 mm Dark Green Colorless Pack of 1000
10541159 20 to 300 μL 55 mm Orange Colorless Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 10708972 Supplier Eppendorf™ Supplier No. 0030000854
Add to basket
Add to basket
Bulk Quote Request
Use these universal pipette tips with a wide variety of pipette models. Eppendorf™ epT.I.P.S.™ Standard Pipettor Tips feature optimal wetting properties, high transparency and special, certified purity grades.
  • Minimal attachment and ejection forces with the highest level of tightness
  • The ergonomically optimized cone geometry provides the optimal pipette fit on the Eppendorf pipette
  • Fine graduations make visual checks of the pipetting volume even easier
  • Fine retracted or extended tips simplify work with small volumes
  • Long, narrow tips reach the base of narrow and high vessels without touching the top of the vessel edge
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 0.5 to 20 μL
Filtered Non-filtered
Color Colorless
Autoclavable Autoclavable
Length (English) 1.8 in.
Length (Metric) 46 mm
Format Bulk
Tip Style Universal
Packaging 1000 Tips (2 Bags x 500 Tips)
Quantity 1000 Pk.
Product Type Pipette Tips
Color Code Light Gray
For Use With (Equipment) Eppendorf, Universal
Graduated Yes
Product Line epT.I.P.S.
Graduations Fine