Learn More
Eppendorf epT.I.P.S.™ Motion Pipette Tips
Shop All Eppendorf Products
Click to view available options
Volume (Metric):
10 μL
50 μL
300 μL
1000 μL
Sterility:
Non-sterile
PCR Clean
Sterile
Filtered:
Filtered
Non-filtered
Unit Size:
Pack of 960
Description
- Specially designed for use on the epMotion™ 5070, 5073 and 5075 Liquid Handling Workstation
- Motion pipette tips, with and without filters, developed in torsion-resistant, clean racks
- High-precision manufacturing guarantees a perfect fit and extremely accurate dispensing behavior
- Optimized for simple automatic ejection in the Liquid Handling Workstation collection vessel
- Can also be integrated into the workstation as a refilling system
- Packaged in contamination-free racks
- Available in the following purity grades: standard Eppendorf Quality, PCR clean, Sterile, PCR clean and Sterile
Specifications
Specifications
| Volume (Metric) | 1000 μL |
| Filtered | Filtered |
| Sterility | PCR Clean |
| Color | Clear |
| Autoclavable | Not Autoclavable |
| Format | Box |
| Tip Style | Standard, Straight |
| Product Type | Robotic Tips |
| Disposable | Single-use |
| For Use With (Equipment) | Eppendorf epMotion |
| Show More |