Learn More
Eppendorf epT.I.P.S.™ Motion as Reload System
GREENER_CHOICE
Product Code. 30019052 Shop All Eppendorf Products
Change view
Click to view available options
Volume (Metric):
0.5 to 50 μL
10 μL
10 to 1000 μL
5 to 300 μL
Sterility:
Non-sterile
PCR Clean
Sterile
Filtered:
Filtered
Non-filtered
Unit Size:
Pack of 2304
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sterility Filtered Unit Size
30019052 0.5 to 50 μL Non-sterile Non-filtered Pack of 2304
30019045 0.5 to 50 μL PCR Clean Filtered Pack of 2304
30019040 0.5 to 50 μL Sterile Filtered Pack of 2304
30019036 5 to 300 μL Non-sterile Non-filtered Pack of 2304
30019038 5 to 300 μL PCR Clean Filtered Pack of 2304
30019042 5 to 300 μL Sterile Filtered Pack of 2304
30019031 10 to 1000 μL Non-sterile Non-filtered Pack of 2304
30019053 10 to 1000 μL PCR Clean Filtered Pack of 2304
30019030 10 μL Non-sterile Non-filtered Pack of 2304
30019046 10 μL PCR Clean Filtered Pack of 2304
30019029 10 μL Sterile Filtered Pack of 2304
30019056 10 to 1000 μL Sterile Filtered Pack of 2304
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 30019052 Supplier Eppendorf Supplier No. 0030014421
In Stock
Add to basket
Edge
Add to basket
Eppendorf epT.I.P.S. Motion work with the epMotion platform, with a TipHolder accessory the reload trays offer an environmentally friendly option.
  • epT.I.P.S. Motion can be used with epMotion™ 96 or integrated into your workstation as a reload system when combined with the tip holder
  • The high-quality reload tips are delivered in a PET thermoformed tray with sealed lid. Upgrading to the autoclavable, aluminum tip holder, which replaces the disposable box, provides waste reduction of up to 40%
  • Eppendorf consumables and thus also epT.I.P.S. Motion pipette tips as Reload System are shipped in packaging consisting of at least 62% recycled cardboard
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 0.5 to 50 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Not Autoclavable
Format Hanging Tip Rack
Tip Style Standard, Straight
Product Type Robotic Tips
Disposable Single-use
For Use With (Equipment) Eppendorf epMotion
Graduated No