Learn More
Eppendorf epT.I.P.S.™ Motion as Reload System
Shop All Eppendorf Products
Click to view available options
Volume (Metric):
0.5 to 50 μL
10 μL
10 to 1000 μL
5 to 300 μL
Sterility:
Non-sterile
PCR Clean
Sterile
Filtered:
Filtered
Non-filtered
missing translation for 'unitSize'
Pack of 2304
Beschreibung
- epT.I.P.S. Motion can be used with epMotion™ 96 or integrated into your workstation as a reload system when combined with the tip holder
- The high-quality reload tips are delivered in a PET thermoformed tray with sealed lid. Upgrading to the autoclavable, aluminum tip holder, which replaces the disposable box, provides waste reduction of up to 40%
- Eppendorf consumables and thus also epT.I.P.S. Motion pipette tips as Reload System are shipped in packaging consisting of at least 62% recycled cardboard
Spezifikation
Spezifikation
| Volume (Metric) | 10 μL |
| Filtered | Filtered |
| Sterility | PCR Clean |
| Color | Clear |
| Autoclavable | Not Autoclavable |
| Format | Hanging Tip Rack |
| Tip Style | Standard, Straight |
| Product Type | Robotic Tips |
| Disposable | Single-use |
| For Use With (Equipment) | Eppendorf epMotion |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur