Description
- epT.I.P.S. Motion can be used with epMotion™ 96 or integrated into your workstation as a reload system when combined with the tip holder
- The high-quality reload tips are delivered in a PET thermoformed tray with sealed lid. Upgrading to the autoclavable, aluminum tip holder, which replaces the disposable box, provides waste reduction of up to 40%
- Eppendorf consumables and thus also epT.I.P.S. Motion pipette tips as Reload System are shipped in packaging consisting of at least 62% recycled cardboard
Specifications
Specifications
| Volume (Metric) | 5 to 300 μL |
| Filtered | Non-filtered |
| Sterility | Non-sterile |
| Color | Clear |
| Autoclavable | Not Autoclavable |
| Format | Hanging Tip Rack |
| Tip Style | Standard, Straight |
| Product Type | Robotic Tips |
| Disposable | Single-use |
| For Use With (Equipment) | Eppendorf epMotion |
| Show More |