Learn More
Eppendorf epT.I.P.S.™ Motion as Reload System
GREENER_CHOICE
Product Code. 30019029 Shop All Eppendorf Products
Change view
Click to view available options
Volume (Metric):
0.5 to 50 μL
10 μL
10 to 1000 μL
5 to 300 μL
Sterility:
Non-sterile
PCR Clean
Sterile
Filtered:
Filtered
Non-filtered
Unit Size:
Pack of 2304
Product Code. Volume (Metric) Sterility Filtered unitSize
30019029 10 μL Sterile Filtered Pack of 2304
30019052 0.5 to 50 μL Non-sterile Non-filtered Pack of 2304
30019045 0.5 to 50 μL PCR Clean Filtered Pack of 2304
30019040 0.5 to 50 μL Sterile Filtered Pack of 2304
30019036 5 to 300 μL Non-sterile Non-filtered Pack of 2304
30019038 5 to 300 μL PCR Clean Filtered Pack of 2304
30019042 5 to 300 μL Sterile Filtered Pack of 2304
30019031 10 to 1000 μL Non-sterile Non-filtered Pack of 2304
30019053 10 to 1000 μL PCR Clean Filtered Pack of 2304
30019030 10 μL Non-sterile Non-filtered Pack of 2304
30019046 10 μL PCR Clean Filtered Pack of 2304
30019056 10 to 1000 μL Sterile Filtered Pack of 2304
12 options
Product Code. 30019029
Brand: Eppendorf 0030014561
Only null left
Add to basket
Add to basket
Eppendorf epT.I.P.S. Motion work with the epMotion platform, with a TipHolder accessory the reload trays offer an environmentally friendly option.
  • epT.I.P.S. Motion can be used with epMotion™ 96 or integrated into your workstation as a reload system when combined with the tip holder
  • The high-quality reload tips are delivered in a PET thermoformed tray with sealed lid. Upgrading to the autoclavable, aluminum tip holder, which replaces the disposable box, provides waste reduction of up to 40%
  • Eppendorf consumables and thus also epT.I.P.S. Motion pipette tips as Reload System are shipped in packaging consisting of at least 62% recycled cardboard
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 10 μL
Filtered Filtered
Sterility Sterile
Color Clear
Autoclavable Not Autoclavable
Format Hanging Tip Rack
Tip Style Standard, Straight
Product Type Robotic Tips
Disposable Single-use
For Use With (Equipment) Eppendorf epMotion
Graduated No