Learn More
Eppendorf™ Xplorer™ plus Electronic Pipette, Multi-Channel Product Code.: 16282401
Eppendorf™ Xplorer™ plus Electronic Pipette, Multi-Channel
GREENER_CHOICE
Product Code. 16282401 Shop All Eppendorf Products
5 to 100 μL, 16, Light Yellow, Each
Click to view available options
Volume (Metric):
0.5 to 10 μL
1 to 20 μL
15 to 300 μL
5 to 100 μL
50 to 1200 μL
Number of Channels:
8
12
16
24
Color:
Green
Light Pink
Light Yellow
Medium Gray
Orange
Yellow
Unit Size:
Each
Product Code. 16282401
Brand: Eppendorf™ 4861000792
Xplorer™ plus Multi-Channel Electronic Pipette with intuitive operating system and design for simplicity, precision and reproducibility.
Facilitates performance of additional, intelligent applications including: aspirating, diluting, sequential dispensing and reverse pipetting. Also, helps eliminate delays due to complicated programming and/or inflexible processes. Provides precisely adjustable parameters, maximum reproducible results, fatigue-free work and consistent, full control over the pipetting processes.
  • Multifunctional rocker: ''Up is up and down is down™'' principle
  • Easy selection of all functions with the practical selection dial
  • Operate in the language of your choice
  • Clearly arranged color display; no sub-menus; help function
  • Innovative rocker: When the tip is ejected, the piston automatically returns to zero position
  • ON/OFF switch: Switches the pipetter off during longer periods of inactivity
  • Additional applications include aspirating, diluting, sequential dispensing and reverse pipetting
  • Programming: One-time programming of sequences for easier handling of complex processes
  • Individual fixed volume setting: Desired volume can be saved
  • Password protection: Protects programming and adjustment
  • Individual speed setting: Sets parameters to your individual working speed
  • Pipetter maintenance reminder function, based on time or frequency of use as desired
Reliably reproducible results
  • Tips always rest in the same position
  • Volume and speed precisely and reproducibly adjustable
  • Adjustable to your work environment and requirements
PhysioCare Concept™
  • Reduced weight
  • Improved ergonomics
  • Perfect balance
  • Optimally angled pipetter head
  • Improved view and high resolution display
  • Spring-loaded tip cone provides maximum tightness
Applications
  • Pipetting liquids (manual and automatic)
  • Dispensing (timed and manual)
  • Pipetting and mixing
Greener Choice Claims
  • Environmental benefits include:
    • Non-Toxic
    • Free-of a substance that causes environmental harm
    • Renewable Energy
    • Ozone-Safe or Safe for the Ozone Layer
TRUSTED_SUSTAINABILITY
Specifications
Battery Type Lithium-ion
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Light Yellow
Disposable Reusable
Number of Channels 16
Product Line Xplorer Plus
Programmability Yes
Type Electronic Pipette
Volume (Metric) 5 to 100 μL
Display LCD
Finger Style Ergonomic
Includes 10 Pipetting modes, pipetting history, password protection, a service indicator and the ability to program multiple modes in a sequence for custom applications
Multilingual User Interface Yes
Product Title
Eppendorf™ Xplorer™ plus Electronic Pipette, Multi-Channel >