Learn More
Eppendorf™ S-4-72 Rotor Adapters
Shop All Eppendorf Products
Click to view available options
Capacity:
1 x 250 mL Tubes
12 x 7/17 mL Tubes
13 x 5.5/12 mL Tubes
14 x 2.6/8 mL Tubes
2 x 50 mL Conical Tubes
26 x 1.5/2 mL Tubes
4 x 50 mL Conical Tubes
8 x 15 mL Conical Tubes
8 x 5 mL Conical Tubes
Unit Size:
Pack of 2
Description
- For use with Centrifuge 5804/5804 R and 5810/5810 R
Specifications
Specifications
| Type | Adapter |
| Tube Type | Microtube |
| Capacity | 26 x 1.5/2 mL Tubes |
| For Use With (Equipment) | S-4-72 (4 x 250mL) rotor in Model 5804/5810 centrifuges |
| Tube Size | 1.5/2 mL |