Learn More
Eppendorf™ Rotor for Centrifuge 5427 R
Product Code. 13463396 Shop All Eppendorf Products
Change view
Click to view available options
Capacity:
12 x 5 mL Eppendorf Tubes
24 Spin Columns
24 x 1.5 to 2.0 mL Tubes
24 x 1.5/2 mL
30 x 1.5 to 2.0 mL Tubes
48 x 0.2 mL PCR Tubes
48 x 1.5 to 2.0 mL Tubes
Includes:
Aerosol-tight with Rotor Lid
Plastic Rotor Lid
Rotor Lid
Unit Size:
Each
Product Code. Capacity Includes unitSize
13463396 30 x 1.5 to 2.0 mL Tubes Rotor Lid Each
13443396 24 x 1.5 to 2.0 mL Tubes Aerosol-tight with Rotor Lid Each
13473396 30 x 1.5 to 2.0 mL Tubes Aerosol-tight with Rotor Lid Each
13453396 48 x 1.5 to 2.0 mL Tubes Aerosol-tight with Rotor Lid Each
15148354 12 x 5 mL Eppendorf Tubes Aerosol-tight with Rotor Lid Each
13483396 24 Spin Columns Aerosol-tight with Rotor Lid Each
13493396 48 x 1.5 to 2.0 mL Tubes Rotor Lid Each
13403406 48 x 0.2 mL PCR Tubes Plastic Rotor Lid Each
13413406 24 x 1.5/2 mL Aerosol-tight with Rotor Lid Each
9 options
Product Code. 13463396
Brand: Eppendorf™ 5409708007
Eppendorf Rotor for Centrifuge 5427 R redefine efficiency. Tailor your centrifugation with various capacities and speeds. Explore accessory options for enhanced versatility, ensuring precise sample processing for diverse applications.
  • Precision-engineered for optimal sample separation.
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
  • Accessories (adaptors and buckets) further customize your centrifuge setup.
  • Aerosol-tight options available.
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor
Capacity 30 x 1.5 to 2.0 mL Tubes
Max. RCF 20,817 x g
Max. Speed 14,000 rpm
For Use With (Equipment) Centrifuge 5427 R
Includes Rotor Lid
No. of Places 30
Model F-45-30-11