Learn More
Eppendorf™ Rotor for Centrifuge 5427 R
Shop All Eppendorf Products
Click to view available options
Capacity:
12 x 5 mL Eppendorf Tubes
24 Spin Columns
24 x 1.5 to 2.0 mL Tubes
24 x 1.5/2 mL
30 x 1.5 to 2.0 mL Tubes
48 x 0.2 mL PCR Tubes
48 x 1.5 to 2.0 mL Tubes
Includes:
Aerosol-tight with Rotor Lid
Plastic Rotor Lid
Rotor Lid
Unit Size:
Each
Description
- Precision-engineered for optimal sample separation.
- All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
- Accessories (adaptors and buckets) further customize your centrifuge setup.
- Aerosol-tight options available.
Specifications
Specifications
| Type | Aerosol-tight Rotor |
| Capacity | 24 x 1.5 to 2.0 mL Tubes |
| Max. RCF | 25,001 x g |
| Max. Speed | 16,220 rpm |
| For Use With (Equipment) | Centrifuge 5427 R |
| Includes | Aerosol-tight with Rotor Lid |
| Model | FA-45-24-11 |
Product Content Correction
Product Title
Eppendorf™ Rotor for Centrifuge 5427 R >