Learn More
Eppendorf™ Research™ plus, Mechanical Single-Channel Pipettes with Fixed Volume
Shop All Eppendorf Products
Click to view available options
Volume (Metric):
10 μL
20 μL
25 μL
50 μL
100 μL
200 μL
250 μL
500 μL
1000 μL
Accuracy:
±0.6%
±0.7%
±0.8%
±1%
±1.2%
Color:
Blue
Gray
Yellow
Unit Size:
Each
Description
- Control button and ejector's low operating force are positioned for ergonomics
- 4-digit, magnifying-shape volume display Calibration display and adjustment opening for specific liquids and volumes Ultra light PerfectPiston™ piston system constructed of chemically resistant Fortron™ Quick connection clip allows removal of lower part in two quick steps (not in 10μL pipetters) Spring loaded tip cone promotes tight fit to the tip Color indicates pipetter's volume
Spécification
Spécification
| Accuracy | ±0.6% |
| Autoclavable | Autoclavable |
| Color | Blue |
| For Use With | 1000 μL Pipette Tip |
| Type | Fixed Volume Pipette |
| Volume (Metric) | 250 μL |
| For Use With (Equipment) | 1000μL Pipette tip |
| Includes | Spring loaded tip cone, ejector, blue operating button |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu