Learn More
Eppendorf™ Research™ plus, Mechanical Single-Channel Pipettes with Fixed Volume
GREENER_CHOICE
Product Code. 15703469 Shop All Eppendorf Products
Click to view available options
Volume (Metric):
10 μL
20 μL
25 μL
50 μL
100 μL
200 μL
250 μL
500 μL
1000 μL
Accuracy:
±0.6%
±0.7%
±0.8%
±1%
±1.2%
Color:
Blue
Gray
Yellow
Unit Size:
Each
Product Code. 15703469
Brand: Eppendorf™ 3124000067
Only null left
Add to basket
Add to basket
Resistant to chemical, thermal, and mechanical stress
  • Control button and ejector's low operating force are positioned for ergonomics
  • 4-digit, magnifying-shape volume display Calibration display and adjustment opening for specific liquids and volumes Ultra light PerfectPiston™ piston system constructed of chemically resistant Fortron™ Quick connection clip allows removal of lower part in two quick steps (not in 10μL pipetters) Spring loaded tip cone promotes tight fit to the tip Color indicates pipetter's volume
TRUSTED_SUSTAINABILITY
Specifications
Accuracy ±0.7%
Autoclavable Autoclavable
Color Yellow
For Use With 200 μL Pipette Tip
Type Fixed Volume Pipette
Volume (Metric) 50 μL
For Use With (Equipment) 200μL Pipette tip
Includes Spring loaded tip cone, ejector, yellow operating button
Product Content Correction
Product Title
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.