Learn More
Eppendorf Reference™ 2, Mechanical Single-Channel Pipette
GREENER_CHOICE
Product Code. 15753479 Shop All Eppendorf Products
Change view
Click to view available options
Volume (Metric):
0.1 to 2.5 μL
0.25 to 2.5 mL
0.5 to 10 μL
0.5 to 5 mL
1 to 10 mL
10 to 100 μL
100 to 1000 μL
2 to 20 μL
20 to 200 μL
30 to 300 μL
Color:
Blue
Dark Gray
Light Gray
Medium Gray
Orange
Red
Turquoise
Violet
Yellow
Increments:
0.002 μL
0.01 μL
0.02 μL
0.1 μL
0.2 μL
1 μL
2 μL
5 μL
10 μL
Unit Size:
Each
Product Code. Volume (Metric) Color Increments unitSize
15753479 2 to 20 μL Light Gray 0.02 μL Each
15743479 0.5 to 10 μL Medium Gray 0.01 μL Each
15733479 0.1 to 2.5 μL Dark Gray 0.002 μL Each
15763479 2 to 20 μL Yellow 0.02 μL Each
15773479 10 to 100 μL Yellow 0.1 μL Each
15783479 20 to 200 μL Yellow 0.2 μL Each
15793479 30 to 300 μL Orange 0.2 μL Each
15703489 100 to 1000 μL Blue 1 μL Each
15723489 0.5 to 5 mL Violet 5 μL Each
15713489 0.25 to 2.5 mL Red 2 μL Each
15733489 1 to 10 mL Turquoise 10 μL Each
11 options
Product Code. 15753479
Brand: Eppendorf 4924000037
Only null left
Add to basket
Add to basket
Minimize risk of aerosol contamination and reduce thumb strain with these variable volume pipettes. Eppendorf™ Reference™ 2 Variable Volume Pipettes are ideal for pipetting and mixing liquids, filling of plates, gels and reaction vessels, and phase extraction and removal of supernatants.
  • Integrated pipetting system with internal RFID technology
  • Serial number is printed on multiple components of pipette
  • Color coded and volume labeled pipette for quick identification of volume size/tip size
  • Round upper part of pipette makes it possible to work in every position
  • Four-digit display for exact, quick and secure volume setting (volume lock included)
  • Spring-loaded tip cone reduces effort during operation/ejection
  • Sturdy upper handle, movable parts and external edges made from stainless-steel make pipette robust
  • High flexibility with ability to remove single cones
  • Ensures continuous identical pipet alignment during work process
  • Fully autoclavable pipet with smooth surface and few rough edges
  • Single-button operation
  • Has Spring-loaded tip cone, movable parts, stainless steel external edges with customizable labeling area
  • Ergonomic handling with reduced operating effort
  • Simple and quick tip ejection with active aerosol reduction
  • Reduce repetitive strain injuries by not having to move your thumb sideways
  • Haptic feedback before tip ejection
  • High precision and accuracy provides reliable pipetting results
  • User friendly secondary adjustment
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Color Light Gray
For Use With Pipette Tip 20 μL
Number of Channels 1
Product Line Reference 2
Type Pipette
Volume (Metric) 2 to 20 μL
For Use With (Equipment) 20 μL pipette tips
Includes epT.I.P.S.™ Box 2.0 with 96 Tips
Increments 0.02 μL